accessibility menu, dialog, popup

UserName

CAS RN 762-42-5

IUPAC Name: 1,4-dimethyl but-2-ynedioate
Synonyms: dimethyl acetylenedicarboxylate
Molecular Weight (g/mol): 142.11
Molecular Formula: C6H6O4
InChi Key: VHILMKFSCRWWIJ-UHFFFAOYSA-N
SMILES: COC(=O)C#CC(=O)OC
Boiling Point 
  • (3)
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (9)
  • (3)
  • (5)
  • (4)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (10)