missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, Anhydrous, Reagent, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18606691
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606691 50 kg Fiber Drum
18606686 25 g Amber Glass Bottle
18606687 125 g Amber Glass Bottle
18606688 500 g Amber Glass Bottle
18606689 2.5 kg Amber Glass Bottle
18606690 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606691 Supplier Spectrum Chemical Mfg Cor Supplier No. S137750KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1377, 7601-89-0, NaClO4

Spectrum™ Chemical Sodium Perchlorate, Anhydrous, Reagent, ACS

Chemical Identifiers

CAS 7601-89-0
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.44
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
IUPAC Name sodium perchlorate
SMILES [Na+].[O-][Cl](=O)(=O)=O

Specifications

pH 6.0 to 8.0
CAS Max % 1
Quantity 50 kg
Percent Purity 98 to 102%
Grade ACS
Packaging Fiber Drum