missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, Solstice

Catalog No. 6001532
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001532 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001532 Supplier Solstice Supplier No. 410241500G
Only null left

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7601-89-0
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.436
MDL Number MFCD00003480
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]

Specifications

Quantity 500 g
Linear Formula NaClO4
UN Number UN1502
Formula Weight 122.44g/mol
Percent Purity ≥98%
Chemical Name or Material Sodium perchlorate
Hazard Statement P220-P305 + P351 + P338
RTECSNumber SC9800000