accessibility menu, dialog, popup

UserName

H3COONH2OHHCl

CAS RN 75748-36-6

IUPAC Name: benzyl 2-amino-3-hydroxybutanoate hydrochloride
Synonyms: benzyl 2-amino-3-hydroxybutanoate hydrochloride
Molecular Weight (g/mol): 245.7
Molecular Formula: C11H16ClNO3
InChi Key: IDZGTFSDZJVMSD-UHFFFAOYNA-N
SMILES: Cl.CC(O)C(N)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results