accessibility menu, dialog, popup

UserName

CAS RN 75706-12-6

IUPAC Name: 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
Synonyms: leflunomide lefunomide [inn-spanish] arava leflunomide (substance) aventis pharma brand of leflunomide leflunomidum SU 101 (pharmaceutical) leflunomida 来氟米特 leflunomide (product)
Molecular Weight (g/mol): 270.21
Molecular Formula: C12H9F3N2O2
InChi Key: VHOGYURTWQBHIL-UHFFFAOYSA-N
SMILES: CC1=C(C=NO1)C(=O)NC1=CC=C(C=C1)C(F)(F)F
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)