missing translation for 'onlineSavingsMsg'
Learn More

Leflunomide 98.0+%, TCI America™

Catalog No. L0250200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0250200MG 200 mg
L02501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0250200MG Supplier TCI America Supplier No. L0250200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75706-12-6
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD00867593
InChI Key VHOGYURTWQBHIL-UHFFFAOYSA-N
Synonym leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn
PubChem CID 3899
ChEBI CHEBI:6402
IUPAC Name 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
SMILES CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F

Specifications

Melting Point 168°C
Color White
Quantity 200 mg
UN Number 2811
Formula Weight 270.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Leflunomide
RTECSNumber NY2354200
TSCA No
Recommended Storage Refrigerator