accessibility menu, dialog, popup

UserName

CAS RN 75705-21-4

IUPAC Name: [4-(aminomethyl)phenyl]boronic acid hydrochloride
Synonyms: 4-(aminomethyl)phenylboronic acid hydrochloride
Molecular Weight (g/mol): 187.43
Molecular Formula: C7H11BClNO2
InChi Key: HUZNRXFJHYNUMV-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results