accessibility menu, dialog, popup

UserName

CAS RN 7554-12-3

H3COOOHOOCH3

CAS RN 7554-12-3

IUPAC Name: 1,4-diethyl 2-hydroxybutanedioate
Synonyms: diethyl malate
Molecular Weight (g/mol): 190.2
Molecular Formula: C8H14O5
InChi Key: VKNUORWMCINMRB-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(O)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results