accessibility menu, dialog, popup

UserName

CAS RN 755038-02-9

H3CONNNNNHONHNCH3OCH3CH3

CAS RN 755038-02-9

IUPAC Name: 4-[(8-cyclopentyl-7-ethyl-5-methyl-6-oxo-5,6,7,8-tetrahydropteridin-2-yl)amino]-3-methoxy-N-(1-methylpiperidin-4-yl)benzamide
Synonyms: 4-[(8-cyclopentyl-7-ethyl-5-methyl-6-oxo-7H-pteridin-2-yl)amino]-3-methoxy-N-(1-methylpiperidin-4-yl)benzamide
Molecular Weight (g/mol): 521.67
Molecular Formula: C28H39N7O3
InChi Key: XQVVPGYIWAGRNI-UHFFFAOYNA-N
SMILES: CCC1N(C2CCCC2)C2=NC(NC3=C(OC)C=C(C=C3)C(=O)NC3CCN(C)CC3)=NC=C2N(C)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results