CAS RN 7542-64-5
IUPAC Name:
2,2-dibromo-1-(4-fluorophenyl)ethan-1-one
Synonyms:
2,2-dibromo-1-(4-fluorophenyl)ethanone
Molecular Weight (g/mol):
295.93
Molecular Formula:
C8H5Br2FO
InChi Key:
XFFGHBAVJPOWDG-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C(=O)C(Br)Br
1
–
1
of
1
results
1
Promotions Available
2,2-Dibromo-4'-fluoroacetophenone, 98%
2,2-Dibromo-4'-fluoroacetophenone, 98%
CAS: 7542-64-5 Molecular Formula: C8H5Br2FO Molecular Weight (g/mol): 295.933 MDL Number: MFCD03791309 InChI Key: XFFGHBAVJPOWDG-UHFFFAOYSA-N Synonym: 2,2-dibromo-1-4-fluorophenyl ethanone,alpha,alpha-dibromo-4-fluoroacetophenone,2,2-dibromo-4'-fluoroacetophenone,4'-fluoro-alpha,alpha-dibromoacetophenone,2,2-dibromo-1-4-fluorophenyl ethan-1-one,2,2-bis bromanyl-1-4-fluorophenyl ethanone PubChem CID: 2783255 IUPAC Name: 2,2-dibromo-1-(4-fluorophenyl)ethanone SMILES: C1=CC(=CC=C1C(=O)C(Br)Br)F