accessibility menu, dialog, popup

UserName

CAS RN 75233-61-3

OONOO

CAS RN 75233-61-3

IUPAC Name: 2-(2-nitroethoxy)oxane
Synonyms: 2-(2-nitroethoxy)oxane
Molecular Weight (g/mol): 175.18
Molecular Formula: C7H13NO4
InChi Key: KQDXRRAVDLIMGM-UHFFFAOYNA-N
SMILES: [O-][N+](=O)CCOC1CCCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results