accessibility menu, dialog, popup

UserName

CAS RN 7511-49-1

IUPAC Name: 4'-bromo-3,5-bis(4-bromophenyl)-1,1'-biphenyl
Synonyms: 4'-bromo-3,5-bis(4-bromophenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 543.1
Molecular Formula: C24H15Br3
InChi Key: HJQRITCAXSBOPC-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)