accessibility menu, dialog, popup

UserName

CAS RN 75088-80-1

IUPAC Name: (5R)-5-hydroxy-4-[(2R,6R)-6-hydroxy-5-[(3E)-4-methyl-6-(2,6,6-trimethylcyclohex-1-en-1-yl)hex-3-en-1-yl]-3,6-dihydro-2H-pyran-2-yl]-2,5-dihydrofuran-2-one
Synonyms: manoalide
Molecular Weight (g/mol): 416.56
Molecular Formula: C25H36O5
InChi Key: FGJIDQWRRLDGDB-CPIXEKRISA-N
SMILES: C\C(CCC1=C(C)CCCC1(C)C)=C/CCC1=CC[C@@H](O[C@H]1O)C1=CC(=O)O[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results