accessibility menu, dialog, popup

UserName

CAS RN 75066-50-1

IUPAC Name: 3-benzyl-1,3-thiazol-3-ium bromide
Synonyms: 3-benzyl-1,3-thiazol-3-ium bromide
Molecular Weight (g/mol): 256.16
Molecular Formula: C10H10BrNS
InChi Key: BVVXTLWKNXVYDS-UHFFFAOYSA-M
SMILES: [Br-].C(C1=CC=CC=C1)[N+]1=CSC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)