CAS RN 75-75-2
Methanesulfonic acid, 99%, extra pure
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 MDL Number: MFCD00007518 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N Synonym: methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
Methanesulfonic acid, 98%
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N Synonym: methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
Methanesulfonic acid, 98+%
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 MDL Number: MFCD00007518 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N Synonym: methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
Methanesulfonic acid, 70% aq. soln.
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 MDL Number: MFCD00007518 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N Synonym: methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
Methanesulfonic acid concentrate 0.1 M CH3SO3H in water (0.1 N), eluent concentrate for IC, MilliporeSigma™ Supelco™
This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.
Methanesulfonic acid, pure, 70% solution in water
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 MDL Number: MFCD00007518 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N Synonym: methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
LiChropur™ Methanesulfonic Acid, For HPLC, 99.5% (T), MilliporeSigma™ Supelco™
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.10 MDL Number: MFCD00007518 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N IUPAC Name: methanesulfonic acid SMILES: CS(O)(=O)=O
Methanesulfonic Acid 99.0+%, TCI America™
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 MDL Number: MFCD00007518 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N Synonym: methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
Methanesulfonic Acid 99.0+%, TCI America™
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 MDL Number: MFCD00007518 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N Synonym: methylsulfonic acid,methanesulphonic acid,methanesulfonicacid,kyselina methansulfonova,methansulfonsaeure,ccris 2783,kyselina methansulfonova czech,ch3so3h,methane sulfonic acid,sulfomethane PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
Methanesulfonic Acid, SPEX CertiPrep™
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.1 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N PubChem CID: 6395 ChEBI: CHEBI:27376 IUPAC Name: methanesulfonic acid SMILES: CS(=O)(=O)O
Methanesulfonic Acid, Technical, 69.5-70.5%, Spectrum™ Chemical
CAS: 75-75-2 Molecular Formula: CH4O3S Molecular Weight (g/mol): 96.10 InChI Key: AFVFQIVMOAPDHO-UHFFFAOYSA-N IUPAC Name: methanesulfonic acid SMILES: CS(O)(=O)=O
Sigma Aldrich Methanesulfonic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.