missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid concentrate 0.1 M CH3SO3H in water (0.1 N), eluent concentrate for IC, MilliporeSigma™ Supelco™

Catalog No. 111015440
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015440 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015440 Supplier Merck Emd Millipore Supplier No. 555171L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Specifications

CAS 75-75-2
Linear Formula CH3 SO3 H
Concentration 0.1 M
MDL Number MFCD00007518
Synonym Methanesulfonic acid solution
Type Organic Standard
Quantity 1 L
Formula Weight 96.11