accessibility menu, dialog, popup

UserName

NH2NFFF

CAS RN 74784-70-6

IUPAC Name: 5-(trifluoromethyl)pyridin-2-amine
Synonyms: 5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 162.12
Molecular Formula: C6H5F3N2
InChi Key: RSGVKIIEIXOMPY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=N1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results