accessibility menu, dialog, popup

UserName

CAS RN 74761-42-5

H3CCH3OOHNHOOCH3

CAS RN 74761-42-5

IUPAC Name: 2-[(methoxycarbonyl)amino]-3-methylbutanoic acid
Synonyms: 2-[(methoxycarbonyl)amino]-3-methylbutanoic acid
Molecular Weight (g/mol): 175.18
Molecular Formula: C7H13NO4
InChi Key: CEFVHPDFGLDQKU-UHFFFAOYNA-N
SMILES: COC(=O)NC(C(C)C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results