accessibility menu, dialog, popup

UserName

CAS RN 7463-51-6

IUPAC Name: 4-bromo-3,5-dimethylphenol
Synonyms: 4-bromo-3,5-dimethylphenol
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: WMUWDPLTTLJNPE-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(C)=C1Br
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (4)
  • (4)
  • (2)
  • (6)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)