missing translation for 'onlineSavingsMsg'
Learn More

Scandium arc-cast pellet, 15.9mm (0.63in) dia

Catalog No. AA45118KF
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45118KF 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45118KF Supplier Thermo Scientific Chemicals Supplier No. 045118KF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-20-2 | Sc | 44.96 g/mol

Scandium arc-cast pellet is used in aerospace industry, oil refineries as a tracing agent, and metal halide lamps/light bulbs. It is mainly used for research purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-20-2
Molecular Formula Sc
Molecular Weight (g/mol) 44.96
MDL Number MFCD00016323
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
Synonym unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
PubChem CID 23952
ChEBI CHEBI:33330
IUPAC Name scandium
SMILES [Sc]

Specifications

Appearance Silver gray pellet
Density 3g/cm
Quantity 1 Pc.
Odor Odorless
Form Pieces
Solubility Information Reacts with water.
Chemical Name or Material Scandium, arc-cast pellet
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 231-129-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only