missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, plasma standard solution, Specpure™ Ru 10,000μg/mL

Catalog No. AA35773AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35773AP 500 mL
AA35773AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35773AP Supplier Thermo Scientific Chemicals Supplier No. 035773AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-18-8 | Ru | 101.07 g/mol

Ruthenium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Density Approx. 1g/cm3 at 20°C
Physical Form Liquid
Solubility Miscible with water
Type Analytical Standard
Quantity 500 mL
Grade Specpure
Chemical Name or Material Ruthenium, Ru 10, 000μg/mL
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation DOT Class: 8; Packing Group: II
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only