missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium cubes, 6mm (0.24in) square, 99.9% (metals basis)

Catalog No. AA45484KF
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45484KF 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45484KF Supplier Thermo Scientific Chemicals Supplier No. 045484KF

missing translation for 'validMainframeMsg'

CAS: 7440-18-8 | Ru | 101.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Quantity 1 Pc.
Odor Odorless
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Ruthenium cubes, 6mm (0.24 in.) square
EINECSNumber 231-127-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only