CAS RN 7391-28-8
IUPAC Name:
3-(4-methylphenyl)-3-oxopropanenitrile
Synonyms:
3-(4-methylphenyl)-3-oxopropanenitrile
Molecular Weight (g/mol):
159.19
Molecular Formula:
C10H9NO
InChi Key:
AIECDYDQPCANJK-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C(=O)CC#N
1
–
1
of
1
results
1
Promotions Available
p-Toluoylacetonitrile, 97%
p-Toluoylacetonitrile, 97%
CAS: 7391-28-8 Molecular Formula: C10H9NO Molecular Weight (g/mol): 159.19 MDL Number: MFCD00067895 InChI Key: AIECDYDQPCANJK-UHFFFAOYSA-N Synonym: 4-methylbenzoylacetonitrile,4-toluoylacetonitrile,3-4-methylphenyl-3-oxopropanenitrile,p-toluoylacetonitrile,3-oxo-3-p-tolyl propanenitrile,3-oxo-3-p-tolylpropanenitrile,acmc-20amdj,pubchem12072,maybridge1_004714,4-methyl-benzoylacetonitrile PubChem CID: 522513 IUPAC Name: 3-(4-methylphenyl)-3-oxopropanenitrile SMILES: CC1=CC=C(C=C1)C(=O)CC#N