accessibility menu, dialog, popup

UserName

CAS RN 7382-59-4

H3COHOOHOOHOCH3

CAS RN 7382-59-4

IUPAC Name: 4-[1-hydroxy-2-(hydroxymethoxy)-2-(2-methoxyphenyl)ethyl]-2-methoxyphenol
Synonyms: 4-[1-hydroxy-2-(hydroxymethoxy)-2-(2-methoxyphenyl)ethyl]-2-methoxyphenol
Molecular Weight (g/mol): 320.34
Molecular Formula: C17H20O6
InChi Key: COXIJIHARCXZBW-UHFFFAOYNA-N
SMILES: COC1=CC(=CC=C1O)C(O)C(OCO)C1=CC=CC=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results