missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015390
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015390 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015390 Supplier Merck Emd Millipore Supplier No. PHR10561G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Trimethoprim is a synthetic antibacterial agent active in vitro against most aerobic gram-negative and gram-positive bacteria. Its mode of action involves inhibition of dihydrofolate reductase, an enzyme involved in catalyzing the reduction of dihydrofolate to tetrahydrofolate in both microbial and eukaryotic cells.

Specifications

CAS 738-70-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C14H18N4O3
MDL Number MFCD00036761
Synonym 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine; NSC 106568
Type Certified Reference Material
Quantity 1 g
Formula Weight 290.32
Grade Certified Reference Material