accessibility menu, dialog, popup

UserName

CAS RN 73590-58-6

IUPAC Name: 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methanesulfinyl]-1H-1,3-benzodiazole
Synonyms: omeprazole ramezol esomeprazole (substance) miracid ortanol zegerid esomeprazole magnesium trihydrate prilosec omeprazole pellets omepradex
Molecular Weight (g/mol): 345.42
Molecular Formula: C17H19N3O3S
InChi Key: SUBDBMMJDZJVOS-UHFFFAOYNA-N
SMILES: COC1=CC=C2N=C(NC2=C1)S(=O)CC1=NC=C(C)C(OC)=C1C
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)