missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015381
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015381 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015381 Supplier Merck Emd Millipore Supplier No. 1932950MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 73590-58-6
Physical Form Neat
Linear Formula C17H19N3O3 S
MDL Number MFCD00083192
Synonym 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1 H-benzimidazole; Antra; Losec
Type Analytical Standard
Quantity 50 mg
Formula Weight 345.42
Percent Purity ≥99% (HPLC)
Grade Analytical Standard