missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015380
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015380 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015380 Supplier Merck Emd Millipore Supplier No. PHR10591G

missing translation for 'validMainframeMsg'

Omeprazole is a benzimidazole drug, which exhibits antisecretory and antiulcer properties. It is widely used in the treatment of duodenal, gastric ulcers, reflux oesophagitis and against other acid-related diseases.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 73590-58-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C17H19N3O3 S
MDL Number MFCD00083192
Synonym 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1 H-benzimidazole; Antra; Losec
Type Certified Reference Material
Quantity 1 g
Formula Weight 345.42
Grade Certified Reference Material