accessibility menu, dialog, popup

UserName

OOHBr(E/Z)

CAS RN 7345-79-1

IUPAC Name: 3-(2-bromophenyl)prop-2-enoic acid
Synonyms: 3-(2-bromophenyl)prop-2-enoic acid
Molecular Weight (g/mol): 227.06
Molecular Formula: C9H7BrO2
InChi Key: OMHDOOAFLCMRFX-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC=CC=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results