accessibility menu, dialog, popup

UserName

CAS RN 7338-27-4

H3COOCH2OHO

CAS RN 7338-27-4

IUPAC Name: 4-methoxy-2-methylidene-4-oxobutanoic acid
Synonyms: 4-methoxy-2-methylidene-4-oxobutanoic acid
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: OIYTYGOUZOARSH-UHFFFAOYSA-N
SMILES: COC(=O)CC(=C)C(O)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results