accessibility menu, dialog, popup

UserName

CH3H3CNONHN

CAS RN 733748-92-0

IUPAC Name: 4-[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]piperidine
Synonyms: 4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidine
Molecular Weight (g/mol): 195.27
Molecular Formula: C10H17N3O
InChi Key: QDRJZNBKSMAEIE-UHFFFAOYSA-N
SMILES: CC(C)C1=NOC(=N1)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results