accessibility menu, dialog, popup

UserName

CAS RN 7327-87-9

IUPAC Name: 1,4-dihydrazinylphthalazine; sulfuric acid
Synonyms: dihydralazine sulfate; sulfuric acid
Molecular Weight (g/mol): 288.28
Molecular Formula: C8H12N6O4S
InChi Key: BWHAMWGGORIDBK-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NNC1=C2C=CC=CC2=C(NN)N=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)