accessibility menu, dialog, popup

UserName

CAS RN 73-03-0

IUPAC Name: (2R,3R,5S)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolan-3-ol
Synonyms: cordycepin
Molecular Weight (g/mol): 251.25
Molecular Formula: C10H13N5O3
InChi Key: OFEZSBMBBKLLBJ-PNFQIUPPNA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1
Percent Purity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)