missing translation for 'onlineSavingsMsg'
Learn More

3'-Deoxyadenosine, 98%

Catalog No. AAJ65366MC
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65366MC 100 mg
AAJ65366MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65366MC Supplier Thermo Scientific Chemicals Supplier No. J65366MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73-03-0 | C10H13N5O3 | 251.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73-03-0
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00037998
InChI Key OFEZSBMBBKLLBJ-PNFQIUPPNA-N
Synonym cordycepin, 3'-deoxyadenosine, cordycepine, 9-cordyceposidoadenine, adenosine, 3'-deoxy, 9-beta-d-3'-deoxyribofuranosyl adenine, unii-gz8vf4m2j8, 9h-purine, 6-amino-9-3-deoxy-beta-d-ribofuranosyl, ccris 3233, gz8vf4m2j8
PubChem CID 6303
ChEBI CHEBI:29014
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)C[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 100 mg
UN Number UN2811
Beilstein 35194
Merck Index 14,2529
IUPAC Name (2R,3R,5S)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolan-3-ol
Formula Weight 251.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3'-Deoxyadenosine
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-791-4
RTECSNumber AU7358610
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only