accessibility menu, dialog, popup

UserName

CAS RN 7281-04-1

IUPAC Name: benzyl(dodecyl)dimethylazanium bromide
Synonyms: benzododecinium bromide dodecyl dimethyl benzyl ammonium bromide
Molecular Weight (g/mol): 384.45
Molecular Formula: C21H38BrN
InChi Key: KHSLHYAUZSPBIU-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results