accessibility menu, dialog, popup

UserName

H3COOHOCH3

CAS RN 728-87-0

IUPAC Name: bis(4-methoxyphenyl)methanol
Synonyms: bis(4-methoxyphenyl)methanol
Molecular Weight (g/mol): 244.29
Molecular Formula: C15H16O3
InChi Key: ZODAOVNETBTTJX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)