accessibility menu, dialog, popup

UserName

CAS RN 72482-64-5

IUPAC Name: 2,4-difluorobenzoyl chloride
Synonyms: 2,4-difluorobenzoyl chloride
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: JSWRVDNTKPAJLB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C(Cl)=O)C(F)=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)