missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. D188210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D188210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D188210G Supplier TCI America Supplier No. D188210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72482-64-5
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.55
MDL Number MFCD00000658
InChI Key JSWRVDNTKPAJLB-UHFFFAOYSA-N
Synonym 2,4-difluorobenzoylchloride, benzoyl chloride, 2,4-difluoro, benzoyl chloride,2,4-difluoro, acmc-1bfet, 2,4difluorobenzoylchloride, 2,4-difluorobezoyl chloride, 2,4 difluorobenzoyl chloride, 2,4-difluoro benzoylchloride, 2,4-difluoro-benzoylchloride, 2,4-difluoro-benzoyl chloride
PubChem CID 588081
IUPAC Name 2,4-difluorobenzoyl chloride
SMILES FC1=CC=C(C(Cl)=O)C(F)=C1

Specifications

Color Yellow
Boiling Point 70°C
Quantity 10 g
UN Number 3265
Formula Weight 176.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzoyl Chloride