missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzoyl chloride, 97%

Catalog No. AAA1792014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1792014 25 g
AAA1792006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1792014 Supplier Thermo Scientific Chemicals Supplier No. A1792014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72482-64-5 | C7H3ClF2O | 176.55 g/mol

2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.

Chemical Identifiers

CAS 72482-64-5
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.55
MDL Number MFCD00000658
InChI Key JSWRVDNTKPAJLB-UHFFFAOYSA-N
PubChem CID 588081
IUPAC Name 2,4-difluorobenzoyl chloride
SMILES FC1=CC=C(C(Cl)=O)C(F)=C1

Specifications

Density 1.437
Boiling Point 96°C to 98°C (15 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.5145
Quantity 25 g
UN Number UN3265
Beilstein 4800566
Sensitivity Moisture sensitive
Synonym 2,4-difluorobenzoylchloride, benzoyl chloride, 2,4-difluoro, benzoyl chloride,2,4-difluoro, acmc-1bfet, 2,4difluorobenzoylchloride, 2,4-difluorobezoyl chloride, 2,4 difluorobenzoyl chloride, 2,4-difluoro benzoylchloride, 2,4-difluoro-benzoylchloride, 2,4-difluoro-benzoyl chloride
Solubility Information Reacts with water.
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 2,4-Difluorobenzoyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 276-690-4
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only