accessibility menu, dialog, popup

UserName

OHONONOClOOO(?)

CAS RN 72102-91-1

IUPAC Name: 16-(2-carboxyphenyl)-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium perchlorate
Synonyms: 16-(2-carboxyphenyl)-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium perchlorate
Molecular Weight (g/mol): 591.06
Molecular Formula: C32H31ClN2O7
InChi Key: DJWWHVKRLDNDJK-UHFFFAOYSA-N
SMILES: [O-][Cl](=O)(=O)=O.OC(=O)C1=CC=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results