accessibility menu, dialog, popup

UserName

CAS RN 72-43-5

IUPAC Name: 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
Synonyms: methoxychlor DMDT dianisyl trichloroethane mezox K caswell no. 550 methoxcide methoxy ddt higalmetox Dimethoxy-DDT epa pesticide chemical code 034001
Molecular Weight (g/mol): 345.64
Molecular Formula: C16H15Cl3O2
InChi Key: IAKOZHOLGAGEJT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(C1=CC=C(OC)C=C1)C(Cl)(Cl)Cl
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results