missing translation for 'onlineSavingsMsg'
Learn More

Methoxychlor, Restek

Catalog No. 06719861
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719861 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719861 Supplier Chromatographic Supplier No. RK 32233

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72-43-5
Molecular Formula C16H15Cl3O2
Molecular Weight (g/mol) 345.644
InChI Key IAKOZHOLGAGEJT-UHFFFAOYSA-N
PubChem CID 4115
ChEBI CHEBI:6842
IUPAC Name 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl

Specifications

Quantity 1 mL
Chemical Name or Material Methoxychlor