accessibility menu, dialog, popup

UserName

CAS RN 7194-84-5

H3CCH3

CAS RN 7194-84-5

IUPAC Name: heptatriacontane
Synonyms: N-heptatriacontane
Molecular Weight (g/mol): 521.02
Molecular Formula: C37H76
InChi Key: PXEZIKSRSYGOED-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results