missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptatriacontane 95.0+%, TCI America™
$363.85 - $1062.69
Chemical Identifiers
| CAS | 7194-84-5 |
|---|---|
| Molecular Formula | C37H76 |
| Molecular Weight (g/mol) | 521.015 |
| MDL Number | MFCD00042945 |
| InChI Key | PXEZIKSRSYGOED-UHFFFAOYSA-N |
| PubChem CID | 23598 |
| IUPAC Name | heptatriacontane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 7194-84-5 | |
| 521.015 | |
| PXEZIKSRSYGOED-UHFFFAOYSA-N | |
| heptatriacontane |
| C37H76 | |
| MFCD00042945 | |
| 23598 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
| 7194-84-5 | |
| White | |
| MFCD00042945 | |
| PXEZIKSRSYGOED-UHFFFAOYSA-N | |
| heptatriacontane | |
| 23598 | |
| ≥95.0% (GC) | |
| Heptatriacontane |
| 77°C | |
| C37H76 | |
| 1 g | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 521.015 | |
| 521.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes