accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 7169-97-3

CH3ONHBrN

CAS RN 7169-97-3

IUPAC Name: N-(5-bromopyridin-2-yl)acetamide
Synonyms: N-(5-bromopyridin-2-yl)acetamide
Molecular Weight (g/mol): 215.05
Molecular Formula: C7H7BrN2O
InChi Key: MJFCOXATGBYERZ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Br)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.