accessibility menu, dialog, popup

UserName

H3COClFFF

CAS RN 71648-45-8

IUPAC Name: 1-[2-chloro-5-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 222.59
Molecular Formula: C9H6ClF3O
InChi Key: YRGBMTWHOFQSDJ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC=C1Cl)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)