accessibility menu, dialog, popup

UserName

CAS RN 7138-40-1

H3COHO

CAS RN 7138-40-1

IUPAC Name: heptacosanoic acid
Synonyms: heptacosanoic acid carbocerate carboceric acid
Molecular Weight (g/mol): 410.73
Molecular Formula: C27H54O2
InChi Key: VXZBFBRLRNDJCS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results