CAS RN 70852-56-1
CAS RN 70852-56-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Pharmalytes™ pH range 5.0 - 8.0, MilliporeSigma™ Supelco™
Pharmalytes™ are ampholytes which are used to create a pH gradient under application of an electric field.
2
Pharmalytes™ pH range 4.0 - 6.5, MilliporeSigma™ Supelco™
Pharmalytes™ are ampholytes which are used to create a pH gradient under application of an electric field.
3
Pharmalytes™ pH range 3.0 - 10.0, MilliporeSigma™ Supelco™
Pharmalytes™ are ampholytes which are used to create a pH gradient under application of an electric field.
4
Pharmalytes™ pH range 2.5 - 5.0, MilliporeSigma™ Supelco™
Pharmalytes™ are ampholytes which are used to create a pH gradient under application of an electric field.