missing translation for 'onlineSavingsMsg'
Learn More

Pharmalytes™ pH range 2.5 - 5.0, MilliporeSigma™ Supelco™

Catalog No. 111015312
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015312 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015312 Supplier Merck Emd Millipore Supplier No. P164725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmalytes™ are ampholytes which are used to create a pH gradient under application of an electric field.

Specifications

CAS 70852-56-1
Product Line Pharmalyte
Quantity 25 mL
Synonym Ampholytes

Pharmalyte is a registered trademark of GE Healthcare