accessibility menu, dialog, popup

UserName

CAS RN 70753-61-6

IUPAC Name: calcium bis(2,3,4-trihydroxybutanoate)
Synonyms: calcium bis(2,3,4-trihydroxybutanoate)
Molecular Weight (g/mol): 310.27
Molecular Formula: C8H14CaO10
InChi Key: ZJXGOFZGZFVRHK-UHFFFAOYNA-L
SMILES: [Ca++].OCC(O)C(O)C([O-])=O.OCC(O)C(O)C([O-])=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)